Tlačiť
Measuring cylinders, tall form, class B, borosilicate glass
Dodávateľ: HIRSCHMANN
2230194 2230154 2230160 2230170 2230175 2230180 2230186 2230190 2230193
612-3063EA 129.16 EUR
612-3063 612-1396 612-1397 612-1398 612-1399 612-1400 612-1509 612-1515 612-1516
Measuring cylinders, tall form, class B, borosilicate glass
Válce Válce odměrné
DURAN®, borosilicate glass, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, with spout
  • Short graduation marks, blue graduations
DIN EN ISO 4788
Certifikácie: DIN EN ISO 4788