Tlačiť
Measuring cylinders, tall form, class A, PMP
Dodávateľ: VITLAB
64704 64804 64904 65004 65204 65304
612-4412EA 10.45 EUR
612-4412 612-4413 612-4414 612-4415 612-4417 612-4418
Measuring cylinders, tall form, class A, PMP
Válce Válce odměrné
PMP, transparent, tall form, class A.
  • Pouring spout, hexagonal base and ring-moulded graduations
  • Accuracy is individually checked and cylinders marked with an ‘H'
  • Supplied with test certificate indicating lot number and year of production (also on the cylinder)
DIN 12681 / ISO 6706
Certifikácie: DIN 12681 / ISO 6706