Tlačiť
Mixing measuring cylinders, tall form, class B, borosilicate glass
Dodávateľ: HIRSCHMANN
2330194 2330160 2330170 2330175 2330180 2330186 2330190 2330193
612-3087EA 232.33 EUR
612-3087 612-3080 612-3081 612-3082 612-3083 612-3084 612-3085 612-3086
Mixing measuring cylinders, tall form, class B, borosilicate glass
Válce Válce odměrné
DURAN®, borosilicate glass, with PE stopper, class B.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, NS ground socket
  • Main point graduations, blue graduations
DIN EN ISO 4788
Certifikácie: DIN EN ISO 4788
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable