Tlačiť
Measuring cylinders, tall form, class A, borosilicate glass
Dodávateľ: HIRSCHMANN
2210194 2210154 2210160 2210170 2210175 2210180 2210186 2210190 2210193
612-1395EA 163.43 EUR
612-1395 612-1385 612-1386 612-1387 612-1388 612-1389 612-1390 612-1391 612-1392
Measuring cylinders, tall form, class A, borosilicate glass
Válce Válce odměrné
DURAN®, borosilicate glass, tall form, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification available via website: (www.hirschmannlab.de)
  • Hexagonal glass base, with spout
  • Amber graduations with ring marks at major graduations
DIN EN ISO 4788
Certifikácie: DIN EN ISO 4788
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable