Tlačiť
Measuring cylinders, Tall form, class A, borosilicate glass, Schellbach Stripe
Dodávateľ: HIRSCHMANN
2220154 2220160 2220170 2220175 2220180 2220186 2220190 2220193 2220194
612-3046EA 24.83 EUR
612-3046 612-3047 612-3048 612-3049 612-3050 612-3051 612-3052 612-3053 612-3054
Measuring cylinders, Tall form, class A, borosilicate glass, Schellbach Stripe
Válce Válce odměrné
DURAN®, borosilicate glass, tall form, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking)
  • with dated batch identification available via website: (www.hirschmannlab.de)
  • Hexagonal glass base, with spout
  • Blue graduations with ring marks at major graduations
DIN EN ISO 4788
Certifikácie: DIN EN ISO 4788