Tlačiť
Measuring cylinders, low form, class B, PP or SAN
Dodávateľ: VITLAB
640941 641941 642941 643941 645941 64491 64291 644941 64591 64391 64091 64191
612-2400EA 3.32 EUR
612-2400 612-2414 612-2416 612-2418 612-2423 612-2419 612-2415 612-2420 612-2422 612-2417 612-3748 612-3749
Measuring cylinders, low form, class B, PP or SAN
Válce Válce odměrné
SAN or PP, translucent, short form, class B.
  • Pouring spout, round base and moulded scale
  • SAN models are not autoclavable
  • PP models are autoclavable