Tlačiť
Measuring cylinders, tall form, class B, borosilicate glass
Dodávateľ: HIRSCHMANN
2260160 2260170 2260175 2260180 2260186 2260190 2260193
612-3073EA 12.3 EUR
612-3073 612-3074 612-3075 612-3076 612-3077 612-3078 612-3079
Measuring cylinders, tall form, class B, borosilicate glass
Válce Válce odměrné
DURAN®, borosilicate glass, tall form, class B.
  • Calibrated to contain 'ln'
  • Removable HDPE base and protective collar, with spout
  • Short graduation marks, blue graduations
DIN EN ISO 4788
Certifikácie: DIN EN ISO 4788