Tlačiť
Reusable half masks, Valuair
Dodávateľ: HONEYWELL SAFETY PRODUCTS
1001575 1001573 1001574 1001576
111-3525EA 69.39 EUR
111-3525 111-3520 111-3524 111-1348
Reusable half masks, Valuair
Polomasky a filtrační respirátory Polomasky filtrační opakovatelně použitelné Polomasky
Reusable half masks.
Economy: Silicone-free half masks, with a texture that combines comfort and exceptional impermeability for facial protection.
Premier: Half mask made from supple silicone for good flexibility, comfort and resistance; perspiration evacuation hole to prevent skin irritation and simplified fastening mechanism.
  • Double-side filter system ensures improved visibility
  • Easy, secure fitting with the 'click-fit' mechanism
Filter connection: Twin bayonet
Economy: 109 g; Premier: 140 g
Filter connection: Twin bayonet
Weight: 109 g (size M)
Certifikácie: EN 140
Objednávacie informácie: Valuair half masks are designed for use with the Sperian range of filter cartridges (111-3540, 111-3528, 111-3529, 111-3530, 111-0247).