Tlačiť
Label dots
Dodávateľ: PRECISION DYNAMICS TIMEM
C-1FA C-2A C-2FA C-3FA C-5A C-7A C-13A C-20A
817-0378EA 36.5 EUR
817-0378 817-1647 817-0379 817-0380 817-1648 817-1649 817-1652 817-1653
Label dots
Štítky
Self adhesive, smudge proof labels. Suitable for labelling samples and documents. Bright colours. Accepts a variety of inks.
  • Oil and water-resistant
  • Temperature resistant from −45 to +121 °C
  • Peels off easily and leaves no residue
Diameter: 19,0 mm