Tlačiť
Tissue grinders, Potter-Elvehjem type
Dodávateľ: DWK LIFE SCIENCES
357969 358028 357966 357974 358033 357971 357979 358038 357976 357984 358043 357981 357989 358048 357986 357994 358053 357991
432-5011EA 385.48 EUR
432-5011 432-5031 432-5010 432-5013 432-5034 432-5012 432-5015 432-5037 432-5014 432-5017 432-5040 432-5016 432-5019 432-5043 432-5018 432-5021 432-5046 432-5020
Tissue grinders, Potter-Elvehjem type
Homogenizátory Homogenizátory na tkáně
Mortar in borosilicate glass, PTFE pestle with stainless steel rod. Serrated PTFE pestle disperses homogenate into mortar cylinder more efficiently. Precisely constructed with completely interchangeable parts.
  • Clearance between pestle and tube is 0.1 to 0.15 mm
  • Stainless steel rod Ø 6.3 mm
  • Can be autoclaved
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifikácie: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).