Tlačiť
Tissue grinders, Tenbroeck type
Dodávateľ: DWK LIFE SCIENCES
357422 357424 357426 357421 357428
432-1276EA 352.19 EUR
432-1276 432-1277 432-5000 432-1275 432-5001
Tissue grinders, Tenbroeck type
Homogenizátory Homogenizátory na tkáně
Borosilicate glass. Tissue grinders with interchangeable precision pestles and tubes for tissues such as liver, intestines, and heart.
  • Hollow handle permits packing with ice
  • Expanded reservoir and pouring lip
  • Clearance between pestle and tube 0,1 - 0,15 mm
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifikácie: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).