Tlačiť
Tissue grinders, Dounce type
Dodávateľ: DWK LIFE SCIENCES
357546 357538 357542 357544
432-1273EA 610.51 EUR
432-1273 432-1270 432-1271 432-1272
Tissue grinders, Dounce type
Homogenizátory Homogenizátory na tkáně
Borosilicate glass. Designed to retain a high percentage of cell nuclei and mitochondria in soft tissues. In operation, the pestle ball is encircled in liquid which avoids heat build up. Friction is reduced and there is reduced heat accumulation.
  • Mortar has large reservoir and pouring lip
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifikácie: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Informace o dodávce: Each mortar is supplied with a 'loose' pestle (0,114 mm ±0,025 mm), and a 'tight' pestle (0,05 mm  ±0,025 mm).