Tlačiť
Embedding rings
Dodávateľ: SIMPORT SCIENTIFIC
M460 M460-3 M460-4 M460-5 M460-6
720-0815EA 170.48 EUR
720-0815 720-0816 720-0817 720-0818 720-0819
Embedding rings
Kazety pro zalévání vzorků Kroužky do kazet
PS. These embedding rings are suitable for holding and identifying tissue sample blocks and fit well in microtome chuck adapters.
  • The etched writing surface on the ring is marked with an identification number and placed on top of the sample block
  • Additional paraffin is poured into the base mould to cement the ring onto the tissue block
  • The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage