Tlačiť
Biopsy processing/embedding cassettes, Histosette® II, M486
Dodávateľ: SIMPORT SCIENTIFIC
M486-11 M486-2 M486-3 M486-4 M486-5 M486-6 M486-9 M486-12 M486-7 M486-8 M486-10
720-0234EA 198.15 EUR
720-0234 720-0236 720-0237 720-0238 720-0239 720-0240 720-0243 720-0235 720-0241 720-0242 720-1500
Biopsy processing/embedding cassettes, Histosette® II, M486
Kazety pro zalévání vzorků Kazety bioptické
Acetal polymer. Designed to hold biopsy specimens. Covers are already removed from cassettes and are packed separately in the case.
  • Convenient for cassette labelling instruments as the covers are already removed
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange