Tlačiť
Tissue processing/embedding cassettes, Histosette® II, M492
Dodávateľ: SIMPORT SCIENTIFIC
M492-2 M492-4 M492-5 M492-6 M492-10 M492-11 M492-12 M492-3 M492-8 M492-9 M492-7
720-0909EA 358.2 EUR
720-0909 720-0911 720-0912 720-0913 720-0906 720-0907 720-0908 720-0910 720-0914 720-0915 720-1528
Tissue processing/embedding cassettes, Histosette® II, M492
Kazety pro zalévání vzorků Kazety tkáňové
Acetal polymer.
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Anterior writing area at 45° angle to enable use with leading cassette labelling instruments