Tlačiť
Biopsy processing/embedding cassettes, Micromesh™ M507
Dodávateľ: SIMPORT SCIENTIFIC
M507-10 M507-11 M507-12 M507-2 M507-3 M507-4 M507-5 M507-6 M507-7 M507-8 M507-9
720-0968EA 523.87 EUR
720-0968 720-0969 720-0970 720-0971 720-0972 720-0973 720-0974 720-0975 720-0976 720-0977 720-0978
Biopsy processing/embedding cassettes, Micromesh™ M507
Kazety pro zalévání vzorků Kazety bioptické
Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.
  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems