Tlačiť
Glove liners, polyester, BCR™
Dodávateľ: BERKSHIRE
BGL2.20R BGL3.20R BGL2.20L BGL3.20L
115-0537EA 1438.03 EUR
115-0537 112-0532 112-0616 112-3344
Glove liners, polyester, BCR™
Rukavice
Continuous-filament knitted, these ambidextrous polyester gloves provide protection and comfort during extended wear. Ideal under latex, PVC, or other outer gloves to absorb perspiration from hands during periods of extended wear.
  • Seamless construction prevents unravelling and release of unwanted particles into critical environments
  • Launderable for repeated use
  • Comfortable half-finger design provides greater dexterity than a full-finger glove liner and has reduced courses in the finger and cuff
  • Full-finger style provides complete protection from irritation caused by latex and PVC gloves