Tlačiť
Chemical resistant gloves, UltraNeo 341
Dodávateľ: MAPA
34341010 34341011 34341148 34341019
112-0669EA 35.81 EUR
112-0669 112-0670 112-0671 112-0672
Chemical resistant gloves, UltraNeo 341
Rukavice
This neoprene glove features a smooth heavyweight rubber coating and provides chemical resistance against acids and aliphatic solvents.
  • Very comfortable high-quality cotton lining
  • Longer working life for heavy-duty work
  • Forearm protection due to long cuffs
  • High mechanical performance
Thickness: 1,45 mm
Colour: Black
EN 388 (3221), EN 374 (ACJKL), EN 407 (X1XXXX); Cat. III
Certifikácie: EN 388:2016 (2121X), EN ISO 374-1 Type A (ACLMNS), EN 374-5, EN 407 (X1XXXX); Cat. III