Tlačiť
Kyselinovzdorné rukavice pro čisté prostory, neopren/nitril, Advantech 517
Dodávateľ: MAPA
34517038 34517036 34517037 34517039 34517030
112-0503EA 82.92 EUR
112-0503 112-0501 112-0502 112-0504 112-0500
Kyselinovzdorné rukavice pro čisté prostory, neopren/nitril, Advantech 517
Rukavice
These powder-free cleanroom gloves are made from Tri-polymer blend (nitrile, neoprene, and natural rubber) which offers excellent resistance and strong chemical protection against a great number of chemical products. Free from silicone or paraffin.
  • Extreme comfort and durability, exceptional chemical and mechanical resistance
  • Resistant to etchants, acids and caustics
  • Embossed texture for good grip of wet and dry objects
  • Wide cuffs enable double-gloving
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Thickness: 0,5 mm
Colour: Natural or orange
Certifikácie: EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. III; AQL 1,5; ASTM D130.
Balenie: 1 pair/bag; 12 pairs/bag