Tlačiť
Clean-process gloves, tri-polymer blend, Trilites 994
Dodávateľ: MAPA
34994426 34994427 34994428 34994429
112-0755EA 49.27 EUR
112-0755 112-0756 112-0757 112-0758
Clean-process gloves, tri-polymer blend, Trilites 994
Rukavice
These ambidextrous, liquid-proof gloves with beaded cuffs are made from a natural rubber latex, neoprene and nitrile blend.
  • Powder-free, textured fingertips
  • Provide durability, comfort, tear resistance and chemical splash protection
  • Low particle count maintained in sealed plastic bags
  • Natural rubber latex proteins levels below detection (Lowry test) reducing risk of becoming sensitised
  • Rolled cuff edge prevents tearing when donning and glove roll down on the hand
Ideal for dry handling and splash protection in controlled environments.
Length: 255 mm
Thickness: 0,15 mm
Colour: Purple
EN 374 (Cat. III), AQL 1,5
Certifikácie: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 (MICRO-ORGANISMS), Cat. III; AQL 1,5.
Balenie: 100 gloves/box.