Tlačiť
Rukavice pro čistý proces, latexové, AccuTech® 91-225 / 91-325
Dodávateľ: Ansell
91-225/6 91-225/6.5 91-225/7 91-225/7.5 91-225/8 91-225/8.5 91-225/9
112-0979EA 782.19 EUR
112-0979 112-3668 112-0981 112-0982 112-0983 112-0984 112-0985
Rukavice pro čistý proces, latexové, AccuTech® 91-225 / 91-325
Rukavice
Hand-specific gloves with straight cuffs for all applications requiring a high level of dexterity and grip. Ideal for pharmaceutical, medical device manufacturing and biotech. The sterile version is suitable for use in ISO 6 (Class 1000) cleanroom environments.
  • Sterile or non sterile, powder-free, bisque fingertips
  • Hand-specific, ergonomically designed for improved dexterity, comfort and long wear
  • Straight cuff for better fit over garment
  • Protein level below 50 µg/gram
EN 374, Cat. III, ASTM D 5712-95; AQL 1,0
Thickness: 0,18 mm (palm) 0,2 mm (fingertips), 0,25 mm (cuffs)
Colour: Natural
EN 374, Cat. III, ASTM D 5712-95; AQL 1,0
Certifikácie: EN ISO 374-1:2016 Type B (LPT), EN ISO 374-5:2016 VIRUS; Cat. III; AQL 1,0
Balenie: Packaged in cleanroom compatible packaging and a lined shipping carton. One pair per poly wallet, sealed in poly envelope; 10 pairs per master polybag; 5 master polybags per case liner; 4 case liners per carton.