Tlačiť
Separating funnels, Gilson
Dodávateľ: LENZ LABORGLAS
04030028 04030037 04030049 04030058 04030070 04030073 04030128 04030137 04030149 04030158 04030170 04030173
527-1501EA 92.66 EUR
527-1501 527-1502 527-1503 527-1504 527-1505 527-0024 527-1506 527-1507 527-1508 527-1509 527-1510 527-0025
Separating funnels, Gilson
Nálevky Nálevky dělící
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Without graduation.