Tlačiť
Dropping funnels, cylindrical, without pressure balance
Dodávateľ: LENZ LABORGLAS
04154914 04155814 04157014 04152811 04152814 04153711 04153714 04152211
271-5114EA 111.51 EUR
271-5114 271-5115 271-5116 271-5102 271-5112 271-5103 271-5113 201-0896
Dropping funnels, cylindrical, without pressure balance
Nálevky Nálevky přikapávací
DURAN®, borosilicate glass, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket. With divisions. Stopcock with thread guard.