Tlačiť
Funnels, drum
Dodávateľ: VITLAB
41794 41894 41994 42393 42294
221-1411EA 10.09 EUR
221-1411 221-1412 221-1413 221-1415 221-1414
Funnels, drum
Nálevky Nálevky filtrační Nálevky sudové
PP or HDPE, translucent.
  • Very good (PP) or good (HDPE) chemical resistance
  • Filter insert made from stainless steel and aluminium is available separately for 200 and 250 mm sizes
  • Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.