Tlačiť
Funnels with long stem, DURAN®
Dodávateľ: DWK LIFE SCIENCES
213533306 213533803 213534105
221-0142EA 32.93 EUR
221-0142 221-0143 221-1181
Funnels with long stem, DURAN®
Nálevky Nálevky filtrační Nálevky k filtraci
DURAN®, borosilicate glass 3,3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls. Cone angle 60°.
DIN ISO 4798 (exept 221-0143 and 221-1181)
Certifikácie: DIN ISO 4798 (except 221-0143 and 221-1181)