Tlačiť
Forceps for cover glasses
Dodávateľ: RSG
615.104 615.105 015.145 015.130 015.105 015.104 415.105 415.104
232-2181EA 21.14 EUR
232-2181 232-2182 232-0196 232-0195 232-0194 232-0193 232-0198 232-0197
Forceps for cover glasses
Pinzety
Stainless steel.
  • For easy, secure handling of cover glasses
  • Can also be used for other laboratory items, such as filter discs
  • Easy to clean