Tlačiť
Forceps
Dodávateľ: RSG
413.115 411.115
232-1981EA 72.2 EUR
232-1981 232-1982
Forceps
Pinzety
Stainless steel, PTFE coated.
  • Available in sharp or blunt design
  • Very good chemical and thermal resistance
  • Smooth surfaces prevent contamination and allow easy cleaning