Tlačiť
Volumetric flasks, class B
Dodávateľ: KARTELL
1484 1485 1486 1487 1488 1489
612-3000EA 19.82 EUR
612-3000 612-3001 612-3002 612-3003 612-3004 612-3005
Volumetric flasks, class B
Baňky Baňky odměrné
PP, translucent, with PE stopper.
  • Narrow neck with no meniscus for easy reading
  • Calibrated gravimetrically at +20 °C
  • Autoclavable, chemically non absorbent, non wetting surface