Tlačiť
Volumetric flasks, class A, white markings, BLAUBRAND®
Dodávateľ: BRAND
37438 37443 37445 37446 37447 37448 37449 37450 37451 37452 37453
612-0854EA 90.25 EUR
612-0854 612-0855 612-0856 612-0857 612-0858 612-0859 612-0860 612-0861 612-2748 612-0862 612-0863
Volumetric flasks, class A, white markings, BLAUBRAND®
Baňky Baňky odměrné
Borosilicate glass 3.3, amber, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
*Široké hrdlo.
Certifikácie: DIN EN ISO 1042.
Objednávacie informácie: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.