Tlačiť
Flasks, flat bottom, narrow or wide neck, with flared rim, DURAN®
Dodávateľ: DWK LIFE SCIENCES
217111708 217112404 217113606 217114405 217115401 217116406 217116809 217117102 217117308 217117608 217118604 217312409 217313602 217314401 217315406 217316308 217316402
214-3105EA 22.14 EUR
214-3105 214-3110 214-3115 214-3120 214-3125 214-3130 214-3135 214-3140 214-3145 214-0144 214-0145 214-3210 214-3215 214-3220 214-3225 214-0147 214-3230
Flasks, flat bottom, narrow or wide neck, with flared rim, DURAN®
Baňky Baňky s plochým dnem
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
* Non-DIN ISO size.
DIN ISO 24450 (wide mouth)
Certifikácie: DIN ISO 1773 (narrow neck)
DIN ISO EN 24450 (wide neck)