Tlačiť
Flasks, iodine with collar and iodine stopper, Quickfit®
Dodávateľ: DWK LIFE SCIENCES
FIB250/3 FIB500/3 FIB250/4 FIB500/4
201-8452EA 133.98 EUR
201-8452 201-8454 BIBBFIB250/4 BIBBFIB500/4
Flasks, iodine with collar and iodine stopper, Quickfit®
Baňky Baňky jodové
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For iodine determination. Flask has ground socket and cup shaped top. Stopper has extended handle which projects well above the liquid seal.
BS 2735 (except 201-8453 and 201-8455)
Certifikácie: BS 2735 (except 201-8453 and 201-8455)
Objednávacie informácie: Flasks and stoppers must be ordered seperately.