Tlačiť
Vacuum filtering flasks, Büchner, with KECK™ assembly set, DURAN®
Dodávateľ: DWK LIFE SCIENCES
211946854 211947353 211948658 211948855 211949157 212042452 212043654 212044453 212045458 212046351
511-0325EA 200.56 EUR
511-0325 511-0326 511-0327 511-0328 511-0329 511-0330 511-0331 511-0332 511-0333 511-0334
Vacuum filtering flasks, Büchner, with KECK™ assembly set, DURAN®
Baňky Baňky filtrační
DURAN® borosilicate glass 3.3, clear.
  • Up to 2000 ml in Erlenmeyer shape, from 3000 ml bottle shape
  • Inclusive assembly set with removable hose connection, screw and seals
  • Fulfils "Equipment and product safety regulations"
  • Glass Type I according to USP, EP and JP
For vacuum use.
DIN 12476 and ISO 6556 (only erlenmeyer shape)
Certifikácie: ISO 6556 (Erlenmeyer shape)
DIN 12475 (bottle shape)