Tlačiť
Shaft assembly kit for Celstir® flasks
Dodávateľ: DWK LIFE SCIENCES
356886 356878 356883 356885 356888 356874 356891 356880 356877
734-3156EA 166.06 EUR
734-3156 734-3152 734-3154 734-3155 734-3157 734-3150 734-3158 734-3153 734-3151
Shaft assembly kit for Celstir® flasks
Míchadla Hřídele míchací
A non-breakable stainless steel shaft to replace the glass shaft which is included with Celstir® flasks. May be retrofitted to existing Celstir® flasks.