Tlačiť
Glass caps for culture media bottles, DURAN®
Dodávateľ: DWK LIFE SCIENCES
214411805 214412904
391-0930EA 26.07 EUR
391-0930 391-0931
Glass caps for culture media bottles, DURAN®
Uzávěry Uzávěry k láhvím
DURAN®, borosilicate glass 3.3, clear.
  • Very good temperature resistance
  • Very good chemical resistance