Tlačiť
Erlenmeyer flasks, narrow neck, with standard socket
Dodávateľ: KARTELL
1460 1461 1462 1463 1464 1465
214-0132EA 7.78 EUR
214-0132 214-0133 214-0134 214-0135 214-0136 214-0175
Erlenmeyer flasks, narrow neck, with standard socket
Baňky Baňky dle Erlenmeyera
PP, transparent.
  • Very good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
With moulded graduations. Autoclavable. Supplied without stoppers.