Tlačiť
Erlenmeyer flasks with baffles, Nalgene®
Dodávateľ: Thermo Fisher Scientific
4110-0250 4110-0500 4110-1000 4110-2000
710-4101EA 112.31 EUR
710-4101 710-4102 710-4103 710-4104
Erlenmeyer flasks with baffles, Nalgene®
Baňky Baňky dle Erlenmeyera
PC, transparent.
  • Thermal resistance from −135 to +135 °C
  • Shatter-proof with high mechanical strength
  • Good chemical resistance, but not suitable for solvents, strong acids or alkaline solutions
With four baffles seamlessly incorporated in the base. Ensures excellent mixing of liquids or microbiological cultures. Growth rates for E. coli and S. cerevisiae are comparable with glass flasks. The 500 and 1000 ml flasks have a straight neck accepting 38 mm standard caps made of rust-proof steel or plastic.