Tlačiť
Personal eyewash
Dodávateľ: ORKLA CARE
7200 7221 720300 725200 7205
118-0001EA 24.71 EUR
118-0001 111-0164 111-0165 118-0153 118-0248
Personal eyewash
Pomůcky první pomoci Sprchy na výplach očí
Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.
  • Quick and easy to open
  • Eye cup protected from dust
  • Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
  • Neutralises the pH value
Expiry date: 4 years if container not opened.
* Bottle holder is supplied without eyewash.