Tlačiť
Dishes, evaporating, half depth
Dodávateľ: MORGAN ADVANCED MATERIALS HALD
L109/0 L109/00 L109/000 L109/1 L109/10 L109/2 L109/3 L109/4 L109/4/0 L109/5 L109/6 L109/6A L109/7B L109/9
216-1233EA 17.85 EUR
216-1233 216-4281 216-1232 216-4282 216-1239 216-1234 216-4283 410-9321 216-1231 216-1235 216-4284 216-1236 216-1237 216-1238
Dishes, evaporating, half depth
Podnosy/Misky/Kelímky Misky odpařovací
Porcelain, glazed.
  • Very good chemical resistance
  • Heat resistant up to +1000 °C
  • Low thermal expansion
With spout and round bottom. White glaze inside and outside.
EN 60672-3 Group C100, Type C110
Certifikácie: EN 60672-3 Group C100, Type C110