Tlačiť
Culture dishes for in vitro fertilisation, Nunc™
Dodávateľ: Thermo Fisher Scientific
150255 150270 150360
391-0109EA 509.03 EUR
391-0109 391-0108 391-0475
Culture dishes for in vitro fertilisation, Nunc™
Misky kultivační Misky pro tkáňové kultury
PS, sterile, flat bottom, with lid. These fully certified dishes are specifically intended for in vitro fertilisation (IVF) use.
  • Facilitates gas exchange with vented lid
  • Produced under ISO 13485 with full lot traceability
  • Certificates of conformity based on a 1-cell stage mouse embryo toxicity test, confirm sterility (SAL 10⁶), non pyrogenicity (LAL test), and that the material has passed USP class VI and MEA test requirements