Tlačiť
Cell culture dishes, Nunc™
Dodávateľ: Thermo Fisher Scientific
150288 150318 150326 150350 153066 169558 174888 174926 168381 172931
734-2040EA 284 EUR
734-2040 734-2041 734-2042 734-2043 734-2045 734-2103 734-2113 734-2114 734-1403 734-2704
Cell culture dishes, Nunc™
Misky kultivační Misky pro tkáňové kultury
PS (except 734-2113*, made of Permanox™), sterile, with lid. A large range of dishes, which have been tissue culture-treated.
  • Optically clear and uniform surface suitable for microscopy
  • 60 mm and 35 mm dishes available with grids for cloning or determination of plating efficiency
  • Nunclon™Δ certified surface treatment
These cell culture/petri dishes are made of polystyrene.
These dishes receive the Thermo Scientific™ Nunclon™ Delta surface treatment, a proprietary surface modification that promotes maximum adhesion for a broad range of cell types.