Tlačiť
Cell culture dishes, Falcon®
Dodávateľ: Corning
353001 353002 353003 353004 353025 353037
734-0005EA 312.47 EUR
734-0005 734-0961 734-0006 734-0007 734-0013 734-0016
Cell culture dishes, Falcon®
Misky kultivační Misky pro tkáňové kultury
PS, sterile, with lid. Designed for cell culture, these dishes have a uniform surface chemistry, created by vacuum gas plasma treatment, which promotes cell attachment. The hydrophilic tissue culture surface contains a variety of negatively charged functional groups that support cell attachment and spreading.
  • Flat and optically clear for distortion-free microscopic visualisation of cells
  • Lids designed for optimal gas exchange
  • Stacking rings allow for easier stacking and handling
  • Cell performance tested to ensure consistent results
The design and frosted rim of the Easy-Grip dish improves handling of small dishes. The ability to pick up a small dish conveniently, without accidentally removing the lid, allows faster working and improves aseptic manipulation.