Tlačiť
Jars with loose enfolding lids, DURAN®
Dodávateľ: DWK LIFE SCIENCES
242083409 242084105 242084508 242085701
216-3304EA 59.28 EUR
216-3304 216-3306 216-3308 216-3312
Jars with loose enfolding lids, DURAN®
Nádoby Nádoby víceúčelové
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Low profile, without knob.