Tlačiť
Desiccator cabinets, Nalgene®
Dodávateľ: Thermo Fisher Scientific
5317-0070 5317-0120 5317-0180
467-0022EA 973.93 EUR
467-0022 467-0023 467-0024
Desiccator cabinets, Nalgene®
Exsikátory Boxy exsikační
These transparent acrylic cabinets with stainless steel latches and neoprene gasket provide dust and moisture free storage. Ideal for storing anhydrous, hygroscopic biological or chemical compounds and electronic assemblies. Removable lipped tray holds desiccant.
  • Can be used on benchtop or in refrigerators and freezers
  • Shelves are removable and adjustable, and are vented to maximise airflow
Objednávacie informácie: Supplied with adjustable shelves. Please note that additional or replacement shelves can not be purchased as optional accessories.