Tlačiť
Desiccators
Dodávateľ: KARTELL
230 550 554
467-2120EA 241.08 EUR
467-2120 467-2115 467-2125
Desiccators
Exsikátory Exsikátory plastové
Transparent polycarbonate cover with polypropylene base. The specially grooved stopper ensures air is readmitted very slowly to avoid disturbing the contents.
  • Implosion proof
  • Will withstand vacuum of 740 mm Hg with loss no greater than 20 mm in 24 hours
  • Includes removable pan for holding drying agent
Informace o dodávce: Supplied with vacuum release tap and O-ring.