Tlačiť
Kyvety jednorázové
Dodávateľ: BRAND
759150 759220 759243 759242 759241 759240 759230 759210 759200 759170 759215 759235
612-5684EA 48.94 EUR
612-5684 612-5688 612-5693 612-5692 612-5691 612-5690 612-5689 612-5687 612-5686 612-5685 612-2505 612-2506
Kyvety jednorázové
Kyvety Kyvety spektrofotometrické
Disposable cuvettes often replace expensive and sensitive cuvettes made from glass and quartz.
  • High chemical resistance for use with polar solvents, acids and caustic solutions
  • Measurements at 260 or 280 nm and in the visible wavelength range
  • Round coloured caps, for microcuvettes only, ensure easy sample identification and provide a tight seal to –20 °C
Suitable for photometric measurements of proteins, DNA and RNA in the UV range.
Balenie: Individually wrapped micro UV cuvettes are free of DNase, DNA and RNase.