Tlačiť
Sample tubes, with screw caps, Nunc™
Dodávateľ: Thermo Fisher Scientific
363282 364238
479-0017EA 272.78 EUR
479-0017 479-0019
Sample tubes, with screw caps, Nunc™
Nádobky na vzorky Nádoby pro obecné použití
PS, clear with white PE screw caps.
  • Good chemical resistance, designed for a high level of mechanical stress
  • Temperature resistant from –10 to +70 °C
  • Can be centrifuged up to 4000×g
Flat base. The screw cap does not have sharp inside edges, so preventing the formation of droplets and with only a low risk of aerosol formation. Ideal for transporting and storing blood, urine etc. Working volume 25 ml.