Tlačiť
Sample containers, with snap-on lids
Dodávateľ: KARTELL
731 732 733 734 735 736 737 730
216-3134EA 51.34 EUR
216-3134 216-3138 216-3140 216-3142 216-3144 216-3143 216-3136 216-0576
Sample containers, with snap-on lids
Nádobky na vzorky Nádoby pro obecné použití
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Light and sturdy design
With attached snap-on lid. The lid snaps easily into place with light pressure. Flat base.