Tlačiť
Sample containers, with snap-on caps, Nalgene®
Dodávateľ: Thermo Fisher Scientific
6250-0005 6250-0012 6250-0018 6250-9028 6250-9050 6250-9075
216-8151EA 17.45 EUR
216-8151 216-8152 216-8153 216-8154 216-8155 216-8156
Sample containers, with snap-on caps, Nalgene®
Nádobky na vzorky Nádoby pro obecné použití
LDPE, translucent.
  • Good chemical resistance
  • Heat resistant from –100 to +80 °C
  • Tight fitting snap-on cover, robust
Suitable as a sample container, beaker for pH measurements, weighing container and many other applications.