Tlačiť
Containers, wide mouth, with screw cap, Corning® Gosselin™
Dodávateľ: Corning
PV125-01 PV30-01 PV500-01 PV75-01
215-1309EA 394.71 EUR
215-1309 215-1311 215-1312 215-1323
Containers, wide mouth, with screw cap, Corning® Gosselin™
PP, opaque white, with screw cap.
  • Excellent shock resistance
  • Very good chemical resistance
Ideal for sampling, storage and transport of solid samples.
Pozor: Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904.