Tlačiť
Tubing connectors, straight
Dodávateľ: BRAND
152505 152510 152605 152615
229-3241EA 31.37 EUR
229-3241 229-3242 229-3251 229-3255
Tubing connectors, straight
Fitinky a konektory
PP or HDPE, translucent.
  • Good mechanical strength and chemical resistance
  • PP tubing connector provides excellent thermal resistance, autoclavable
  • HDPE tubing connector provides good thermal resistance
With tapered ends, for connecting tubing with varying diameters.